Amlodipine ≥98%, Moligand™
Supplier: ALADDIN SCIENTIFIC
|
Danger
|
Synonyms:
3-Ethyl 5-methyl 2-((2-aminoethoxy)methyl)-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate
A123260-100G
A123260-1G
A123260-25G
A123260-5G
ALA123260-100GEA
3975.66
USD
ALA123260-100G
ALA123260-1G
ALA123260-25G
ALA123260-5G
Amlodipine ≥98%, Moligand™
Amlodipine
A dihydropyridine calcium channel blocker; activity resides mainly in the (-)-isomer.
Formula:
C₂₀H₂₅ClN₂O₅ Melting Pt: 178-179 ºC Storage Temperature: Freezer |
MDL Number:
MFCD00864687 CAS Number: 88150-42-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...