4-Amino-1-methylpiperidine ≥97% (by GC)

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

A123220-100G A123220-1G A123220-25G A123220-5G
ALA123220-100GEA 165.21 USD
ALA123220-100G ALA123220-1G ALA123220-25G ALA123220-5G
4-Amino-1-methylpiperidine ≥97% (by GC)
4-Amino-1-methylpiperidine
Formula: C₆H₁₄N₂
MW: 114.1888 g/mol
Boiling Pt: 83°C
Density: 0.91 g/cm³
Flash Pt: 39 °C
Storage Temperature: Ambient
MDL Number: MFCD01570549
CAS Number: 41838-46-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=00b872a8655a52656a451f0d34e16a5e94291190&vid=00b872a8655a52656a451f0d34e16a5e94291190&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR