7-Amino-4-(trifluoromethyl)coumarin ≥99%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

A123204-1G A123204-250MG A123204-25G A123204-5G
ALA123204-1GEA 86.9 USD
ALA123204-1G ALA123204250MG ALA123204-25G ALA123204-5G
7-Amino-4-(trifluoromethyl)coumarin ≥99%
7-Amino-4-(trifluoromethyl)coumarin

Product Application:7-Amino-4-(trifluoromethyl)coumarin is used in the synthesis of substrates for the fluorometric assay of proteolytic enzymes in biological fluids. It is suitable as laser dye.

Formula: C₁₀H₆F₃NO₂
Melting Pt: 222-223°C
MDL Number: MFCD00006858
CAS Number: 53518-15-3
EINECS: 258-599-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=532d6f4ead6c01c81965dfd19bb38d5578c37a5b&vid=532d6f4ead6c01c81965dfd19bb38d5578c37a5b&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR