2-Aminopyrimidine-5-carbaldehyde ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A123166-1G A123166-250MG
ALA123166-1GEA 139.56 USD
ALA123166-1G ALA123166250MG
2-Aminopyrimidine-5-carbaldehyde ≥97%
2-Aminopyrimidine-5-carbaldehyde
Formula: C₅H₅N₃O
Melting Pt: 209-214°C
MDL Number: MFCD03701714
CAS Number: 120747-84-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d307f4e27286d6df66f01fc96afa97dd37e17a43&vid=d307f4e27286d6df66f01fc96afa97dd37e17a43&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR