2-(3-Aminophenyl)acetic acid ≥97%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
A123133-1G
A123133-25G
A123133-5G
ALA123133-1GEA
39.81
USD
ALA123133-1G
ALA123133-25G
ALA123133-5G
2-(3-Aminophenyl)acetic acid ≥97%
2-(3-Aminophenyl)acetic acid
Formula:
C₈H₉NO₂ Melting Pt: 147-150°C Storage Temperature: Ambient |
MDL Number:
MFCD00075058 CAS Number: 14338-36-4 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...