5-Aminopicolinic acid ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 5-Aminopyridine-2-carboxylic acid, 5-Amino-2-pyridinecarboxylic acid

A123111-100G A123111-10G A123111-1G A123111-25G A123111-5G
ALA123111-100GEA 1395.46 USD
ALA123111-100G ALA123111-10G ALA123111-1G ALA123111-25G ALA123111-5G
5-Aminopicolinic acid ≥98%
5-Aminopicolinic acid
Formula: C₆H₆N₂O₂
MW: 138.13
Melting Pt: 217-219°C
Storage Temperature: Ambient
MDL Number: MFCD02684600
CAS Number: 24242-20-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c42db12b909977638acc21d9d4d745e14d64632f&vid=c42db12b909977638acc21d9d4d745e14d64632f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR