Adenosine-3',5'-cyclic monophosphate sodium salt ≥99%

Supplier: ALADDIN SCIENTIFIC

A122959-100MG A122959-500MG
ALA122959100MGEA 53.11 USD
ALA122959100MG ALA122959500MG
Adenosine-3',5'-cyclic monophosphate sodium salt ≥99%
Adenosine-3',5'-cyclic monophosphate sodium salt

Adenosine 3′,5′-cyclic monophosphate sodium salt is a naturally-occurring activator of PKA (cAMP-dependent protein kinase A). Cyclic-AMP is an important second messenger that is linked in many systems to neurotransmitter- or hormone-induced receptor stimulation. The cAMP/PKA signaling pathway has shown to induce differentiation, inhibit cell proliferation, and lead to apoptosis. Adenosine 3′,5′-cyclic monophosphate sodium salt monohydrate is an activator of Epac.Adenosine-3′,5′-cyclic monophosphate (cAMP) is an activator of cyclic-AMP-dependent protein kinase A (PKA). The cAMP/PKA signaling pathway has been shown to inhibit cell proliferation, induce differentiation and lead to apoptosis. Exogneous cAMP plays a role in the spontaneous activity and contraction of the heart muscle.

Formula: C₁₀H₁₁N₅NaO₆P
Storage Temperature: Freezer
MDL Number: MFCD00069736
CAS Number: 37839-81-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=23cd5a9c51e2840f30675af6409d69b156d03a36&vid=23cd5a9c51e2840f30675af6409d69b156d03a36&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR