2-Amino-2'-fluoro-2'-deoxyadenosine ≥99%

Supplier: ALADDIN SCIENTIFIC

A122908-1G A122908-250MG
ALA122908-1GEA 277.31 USD
ALA122908-1G ALA122908250MG
2-Amino-2'-fluoro-2'-deoxyadenosine ≥99%
2-Amino-2'-fluoro-2'-deoxyadenosine
Formula: C₁₀H₁₃FN₆O₃
Storage Temperature: Refrigerator
MDL Number: MFCD00900994
CAS Number: 134444-47-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3be68a7f8b400d6ec484f8eb67d750e80bdf384b&vid=3be68a7f8b400d6ec484f8eb67d750e80bdf384b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR