5-Amino-2-fluorobenzonitrile ≥97%

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 3-Cyano-4-fluoroaniline

A122719-100G A122719-25G A122719-5G
ALA122719-100GEA 316.26 USD
ALA122719-100G ALA122719-25G ALA122719-5G
5-Amino-2-fluorobenzonitrile ≥97%
5-Amino-2-fluorobenzonitrile
Formula: C₇H₅FN₂
Melting Pt: 92-96°C
Storage Temperature: Refrigerator
MDL Number: MFCD00277872
CAS Number: 53312-81-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=49e876e8add6cfc70922e848cef1f9de9c4c230b&vid=49e876e8add6cfc70922e848cef1f9de9c4c230b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR