4-Amino-3-fluorobenzonitrile ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

A122718-1G A122718-25G A122718-5G
ALA122718-1GEA 23.66 USD
ALA122718-1G ALA122718-25G ALA122718-5G
4-Amino-3-fluorobenzonitrile ≥98%
4-Amino-3-fluorobenzonitrile
Formula: C₇H₅FN₂
MW: 136.13 g/mol
Melting Pt: 87-90°C
MDL Number: MFCD00055559
CAS Number: 63069-50-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ed6e22f218b56c573ad2f95506bde99ea7fd05db&vid=ed6e22f218b56c573ad2f95506bde99ea7fd05db&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR