2-Amino-4,5-dimethoxybenzonitrile ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A122707-100G A122707-1G A122707-25G A122707-5G
ALA122707-100GEA 3168.16 USD
ALA122707-100G ALA122707-1G ALA122707-25G ALA122707-5G
2-Amino-4,5-dimethoxybenzonitrile ≥98%
2-Amino-4,5-dimethoxybenzonitrile
Formula: C₉H₁₀N₂O₂
Melting Pt: 97-101°C
MDL Number: MFCD00058682
CAS Number: 26961-27-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7994e7bd13eee5cbb5f254268f8157ffff763fbf&vid=7994e7bd13eee5cbb5f254268f8157ffff763fbf&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR