4-Amino-3-chlorobenzonitrile ≥98%

Supplier: ALADDIN SCIENTIFIC
Danger

A122706-100G A122706-25G A122706-5G
ALA122706-100GEA 861.56 USD
ALA122706-100G ALA122706-25G ALA122706-5G
4-Amino-3-chlorobenzonitrile ≥98%
4-Amino-3-chlorobenzonitrile
Formula: C₇H₅ClN₂
Melting Pt: 104 °C
MDL Number: MFCD00052917
CAS Number: 21803-75-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=10bdfab659f458503c85fba53bf68f86dd320c2a&vid=10bdfab659f458503c85fba53bf68f86dd320c2a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR