4-Hydroxy-m-anisidine ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 4-Amino-2-methoxyphenol, 4-Aminoguaiacol, 4-Hydroxy-3-methoxyaniline

A122694-1G A122694-250MG A122694-5G
ALA122694-1GEA 15.11 USD
ALA122694-1G ALA122694250MG ALA122694-5G
4-Hydroxy-m-anisidine ≥98%
4-Hydroxy-m-anisidine
Formula: C₇H₉NO₂
MW: 139.1500 g/mol
Melting Pt: 180 °C
MDL Number: MFCD01707809
CAS Number: 52200-90-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f84cbc0c52db9b6917912a7bcea9ad83c653adfa&vid=f84cbc0c52db9b6917912a7bcea9ad83c653adfa&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR