2-Aminooxazole ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: Oxazol-2-amine

A122648-10G A122648-1G A122648-25G A122648-5G
ALA122648-10GEA 596.51 USD
ALA122648-10G ALA122648-1G ALA122648-25G ALA122648-5G
2-Aminooxazole ≥97%
2-Aminooxazole
Formula: C₃H₄N₂O
MW: 84.10 g/mol
Melting Pt: 90-95°C
Storage Temperature: Refrigerator
MDL Number: MFCD07364485
CAS Number: 4570-45-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=63d933b635d2385d04135bbb8cb26f23660a5d77&vid=63d933b635d2385d04135bbb8cb26f23660a5d77&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR