4-Amino-3-fluorophenol ≥98%

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 2-Fluoro-4-hydroxyaniline

A122547-100G A122547-1G A122547-250MG A122547-25G A122547-5G
ALA122547-100GEA 1899.91 USD
ALA122547-100G ALA122547-1G ALA122547250MG ALA122547-25G ALA122547-5G
4-Amino-3-fluorophenol ≥98%
4-Amino-3-fluorophenol
Formula: C₆H₆FNO
MW: 127.12 g/mol
Melting Pt: 137-140°C
Storage Temperature: Freezer
MDL Number: MFCD00077452
CAS Number: 399-95-1
EINECS: 402-230-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d196b522121e4e1f7b037b614333cdf1570b6dcd&vid=d196b522121e4e1f7b037b614333cdf1570b6dcd&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR