2-Amino-5-fluorophenol ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

A122546-100G A122546-1G A122546-250MG A122546-25G A122546-5G
ALA122546-100GEA 1130.41 USD
ALA122546-100G ALA122546-1G ALA122546250MG ALA122546-25G ALA122546-5G
2-Amino-5-fluorophenol ≥97%
2-Amino-5-fluorophenol
Formula: C₆H₆FNO
MW: 127.12 g/mol
Melting Pt: 135-140°C
Storage Temperature: Refrigerator
MDL Number: MFCD00671759
CAS Number: 53981-24-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9916788c2d5366c8c87a3c2d737dcee5f8eafe87&vid=9916788c2d5366c8c87a3c2d737dcee5f8eafe87&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR