Aztreonam ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: 2-[[(Z)-[1-(2-Amino-4-thiazolyl)-2-[[(2S,3S)-2-methyl-4-oxo-1-sulpho-3-azetidinyl]amino]-2-oxoethylidene]amino]oxy]-2-methylpropionic Acid, 2-((((Z)-1-(2-Aminothiazol-4-yl)-2-(((2S,3S)-2-methyl-4-oxo-1-sulphoazetidin-3-yl)amino)-2-oxoethylidene)amino)oxy)-2-methylpropanoic acid

A122451-100G A122451-1G A122451-25G A122451-5G
ALA122451-100GEA 3263.16 USD
ALA122451-100G ALA122451-1G ALA122451-25G ALA122451-5G
Aztreonam ≥98%
Aztreonam
Formula: C₁₃H₁₇N₅O₈S₂
MW: 435.43 g/mol
Melting Pt: 227°C
Storage Temperature: Freezer
MDL Number: MFCD00072145
CAS Number: 78110-38-0
EINECS: 278-839-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d51444b7cab25fef1afdefc1ef78506593f6b405&vid=d51444b7cab25fef1afdefc1ef78506593f6b405&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR