4-Amino-m-anisic acid ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 4-Amino-3-methoxybenzenecarboxylic acid

A122444-1G A122444-25G A122444-5G
ALA122444-1GEA 11.31 USD
ALA122444-1G ALA122444-25G ALA122444-5G
4-Amino-m-anisic acid ≥98%
4-Amino-m-anisic acid

Key building block for the efficient synthesis of carbazole alkaloids mukonine and mukonidine via oxidative annulation.

Formula: C₈H₉NO₃
MW: 167.16 g/mol
Melting Pt: 184°C
MDL Number: MFCD00016539
CAS Number: 2486-69-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e99824b99129c0e7793f8e94ec71ed82b68b6943&vid=e99824b99129c0e7793f8e94ec71ed82b68b6943&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR