2-Amino-5-chloropyrazine ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 2-amino-5-chloropyrazine

A122400-1G A122400-25G A122400-5G
ALA122400-1GEA 9.41 USD
ALA122400-1G ALA122400-25G ALA122400-5G
2-Amino-5-chloropyrazine ≥98%
2-Amino-5-chloropyrazine
Formula: C₄H₄ClN₃
MW: 129.55 g/mol
Melting Pt: 134 °C
MDL Number: MFCD03423596
CAS Number: 33332-29-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1e6bbbe9e43077c8522d9868b6a70994eb9ce95c&vid=1e6bbbe9e43077c8522d9868b6a70994eb9ce95c&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR