3-Amino-4-methylpyridine ≥98%

Supplier: ALADDIN SCIENTIFIC
Danger

A122386-100G A122386-1G A122386-25G A122386-5G
ALA122386-100GEA 672.51 USD
ALA122386-100G ALA122386-1G ALA122386-25G ALA122386-5G
3-Amino-4-methylpyridine ≥98%
3-Amino-4-methylpyridine
Formula: C₆H₈N₂
MW: 108.14
Boiling Pt: 254 °C
Melting Pt: 102-106°C
Storage Temperature: Ambient
MDL Number: MFCD00128871
CAS Number: 3430-27-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ffd5eba082957a6d94bda993b6c3ccc4eadbb7e&vid=1ffd5eba082957a6d94bda993b6c3ccc4eadbb7e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR