Azilsartan ≥98% (by HPLC), Moligand™
Supplier: ALADDIN SCIENTIFIC
|
Danger
|
A122333-50MG
A122333-5G
A122333-10MG
A122333-1G
A122333-250MG
ALA122333-50MGEA
123.41
USD
ALA122333-50MG
ALA122333-5G
ALA122333-10MG
ALA122333-1G
ALA122333250MG
Azilsartan ≥98% (by HPLC), Moligand™
Azilsartan
Formula:
C₂₅H₂₀N₄O₅ Melting Pt: 188 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD20278186 CAS Number: 147403-03-0 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...