Azido-PEG8-alcohol ≥95%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: O-(2-Azidoethyl)heptaethylene glycol, Azido-dPEG®8-alcohol, Azido-PEG8-OH, N3-PEG8-OH, Azido-PEG8-alcohol, 23-Azido-3,6,9,12,15,18,21-heptaoxatricosan-1-ol, Azido-PEG (n=7)

A122207-100MG A122207-250MG
ALA122207100MGEA 144.31 USD
ALA122207100MG ALA122207250MG
Azido-PEG8-alcohol ≥95%
Azido-PEG8-alcohol
Formula: C₁₆H₃₃N₃O₈
MW: 395.45 g/mol
Storage Temperature: Freezer
CAS Number: 352439-36-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eef2d1dd8633ceeb86eae02bccfa8eabea5a3e0c&vid=eef2d1dd8633ceeb86eae02bccfa8eabea5a3e0c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR