Azido-PEG5-amine ≥95%

Supplier: ALADDIN SCIENTIFIC

A122193-1G A122193-250MG A122193-5G
ALA122193-1GEA 82.56 USD
ALA122193-1G ALA122193250MG ALA122193-5G
Azido-PEG5-amine ≥95%
Azido-PEG5-amine
Formula: C₁₂H₂₆N₄O₅
Density: 1.10 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD16619318
CAS Number: 516493-93-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0d3ee7a58e86796f257f3e47374e728e1b3e3586&vid=0d3ee7a58e86796f257f3e47374e728e1b3e3586&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR