1-Amino-3,6,9,12,15,18-hexaoxahenicosan-21-oic acid ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: Amino-PEG6-carboxylate, 21-Amino-4,7,10,13,16,19-hexaoxahenicosanoic acid, Amino-PEG6-carboxylic acid, NH2-PEG6-CH2CH2COOH, NH2-PEG6-COOH, 1-Amino-3,6,9,12,15,18-hexaoxahenicosan-21-oic acid, Carboxy-PEG6-amine, 21-Amino-4,7,10,13,16,19-hexaoxahenicosanoic Acid

A122171-100MG A122171-1G A122171-250MG
ALA122171100MGEA 154.76 USD
ALA122171100MG ALA122171-1G ALA122171250MG
1-Amino-3,6,9,12,15,18-hexaoxahenicosan-21-oic acid ≥98%
1-Amino-3,6,9,12,15,18-hexaoxahenicosan-21-oic acid

Product Describtion:Amino-PEG6-carboxylic Acid is a novel amino acid that can be manipulated, modified, and used like most other amino acids. The single molecular weight PEG spacer arms between the terminal amine and the terminal carboxylic acid are of exact length (25.1 Å), allowing for precise spatial control, and they are hydrophilic and non-immunogenic, imparting greater water solubility and hydrodynamic volume to target molecules.

Formula: C₁₅H₃₁NO₈
MW: 353.41 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13184943
CAS Number: 905954-28-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a0d40bffe497830a1437de1b1682b141ba743588&vid=a0d40bffe497830a1437de1b1682b141ba743588&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR