Amino-PEG2-acid ≥98%
Supplier: ALADDIN SCIENTIFIC CORPORATION
Synonyms:
Amino-PEG2-C2-acid, α-amine-ω-propionic acid diethylene glycol, H2N-PEG2-CH2CH2COOH, 3-(2-(2-Aminoethoxy)ethoxy)propanoic acid, 3-[2-(2-Aminoethoxy)ethoxy]-propanoic acid
A122168-100MG
A122168-1G
A122168-250MG
A122168-5G
ALA122168100MGEA
112.01
USD
ALA122168100MG
ALA122168-1G
ALA122168250MG
ALA122168-5G
Amino-PEG2-acid ≥98%
Amino-PEG2-acid
Formula:
C₇H₁₅NO₄ MW: 177.198 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD20926377 CAS Number: 791028-27-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...