3-(Carbamimidoylthio)-1-propanesulfonic acid ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: 3-(Carbamimidoylthio)propane-1-sulphonic acid, 3-(Amidinothio)-1-propanesulphonic acid

A121968-100G A121968-25G
ALA121968-100GEA 209.86 USD
ALA121968-100G ALA121968-25G
3-(Carbamimidoylthio)-1-propanesulfonic acid ≥97%
3-(Carbamimidoylthio)-1-propanesulphonic acid
Formula: C₄H₁₀N₂O₃S₂
MW: 198.26 g/mol
Melting Pt: 254-256°C
MDL Number: MFCD00085942
CAS Number: 21668-81-5
EINECS: 244-520-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cd1338bcdfd62233da43182c5f109364c6fbce3e&vid=cd1338bcdfd62233da43182c5f109364c6fbce3e&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR