Azur II eosinate indicator

Supplier: ALADDIN SCIENTIFIC
Danger

A121936-100G A121936-25G
ALA121936-100GEA 978.41 USD
ALA121936-100G ALA121936-25G
Azur II eosinate indicator
Azur II eosinate
Formula: C₅₁H₄₀Br₄Cl₂N₆Na₂O₅S₂
MDL Number: MFCD00084753
CAS Number: 53092-85-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR