Biebrich scarlet Dye content ~60 %

Supplier: ALADDIN SCIENTIFIC
Warning

A121933-100G A121933-25G
ALA121933-100GEA 104.41 USD
ALA121933-100G ALA121933-25G
Biebrich scarlet Dye content ~60 %
Biebrich scarlet
Formula: C₂₂H₁₄N₄Na₂O₇S₂
Melting Pt: 181-188°C
Flash Pt: 300°C
MDL Number: MFCD00003891
CAS Number: 4196-99-0
EINECS: 224-084-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ca6f9c04b3635063b6ff1b0f023d050c15acc9e&vid=4ca6f9c04b3635063b6ff1b0f023d050c15acc9e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR