4-Amino-5-bromo-2-chloropyrimidine ≥95%

Supplier: ALADDIN SCIENTIFIC
Warning

A121855-1G A121855-25G A121855-5G
ALA121855-1GEA 24.61 USD
ALA121855-1G ALA121855-25G ALA121855-5G
4-Amino-5-bromo-2-chloropyrimidine ≥95%
4-Amino-5-bromo-2-chloropyrimidine
Formula: C₄H₃BrClN₃
MW: 208.44 g/mol
MDL Number: MFCD07782040
CAS Number: 205672-25-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d7193af86ed66b314a9a15353d9919b8533a58e4&vid=d7193af86ed66b314a9a15353d9919b8533a58e4&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR