3-Amino-2,6-dimethylpyridine ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 2,6-Dimethylpyridin-3-amine

A121811-100G A121811-10G A121811-1G A121811-25G A121811-5G
ALA121811-100GEA 1974.01 USD
ALA121811-100G ALA121811-10G ALA121811-1G ALA121811-25G ALA121811-5G
3-Amino-2,6-dimethylpyridine ≥97%
3-Amino-2,6-dimethylpyridine
Formula: C₇H₁₀N₂
MW: 122.17 g/mol
Boiling Pt: 230°C
Melting Pt: 121-123°C
MDL Number: MFCD01646409
CAS Number: 3430-33-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=388ce9a5414127ac8e9c639fa6817426e6762618&vid=388ce9a5414127ac8e9c639fa6817426e6762618&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR