3-Amino-1-adamantanol ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 3-Aminoadamantan-1-ol

A121784-100G A121784-25G A121784-5G
ALA121784-100GEA 511.96 USD
ALA121784-100G ALA121784-25G ALA121784-5G
3-Amino-1-adamantanol ≥98%
3-Amino-1-adamantanol

Product Application:It is employed in the synthesis of inhibitors with antihyperglycemic properties. vil- dagliptin is synthesised from 3-Amino-1-adamantanol.

Formula: C₁₀H₁₇NO
MW: 167.25 g/mol
Melting Pt: 265°C
Flash Pt: 110 °C
MDL Number: MFCD01821204
CAS Number: 702-82-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=71e551010369ac686870cfc62125f917b6e1615a&vid=71e551010369ac686870cfc62125f917b6e1615a&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR