1,3-Adamantanediol ≥99%
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Warning
|
Synonyms:
1,3-Dihydroxyadamantane
A121783-5G
A121783-100G
A121783-25G
ALA121783-5GEA
29.36
USD
ALA121783-5G
ALA121783-100G
ALA121783-25G
1,3-Adamantanediol ≥99%
1,3-Adamantanediol
Formula:
C₁₀H₁₆O₂ MW: 168.24 g/mol Boiling Pt: 320.4±10.0 °C Melting Pt: 258°C Density: 1 g/cm³ Flash Pt: 158.5±13.6 °C Storage Temperature: Ambient |
MDL Number:
MFCD00154013 CAS Number: 5001-18-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...