3-Amino-4-hydroxyphenyl methyl sulfone ≥97%

Supplier: Aladdin Scientific
Warning

A121727-25G A121727-5G
ALA121727-25GEA 61.66 USD
ALA121727-25G ALA121727-5G
3-Amino-4-hydroxyphenyl methyl sulfone ≥97%
3-Amino-4-hydroxyphenyl methyl sulfone
Formula: C₇H₉NO₃S
Melting Pt: 158°C
MDL Number: MFCD00719450
CAS Number: 98-30-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=23ee551f8a6e86ffe3d16a36b52a87a271d9a0bd&vid=23ee551f8a6e86ffe3d16a36b52a87a271d9a0bd&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR