2-Amino-6-methylphenol 92%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
6-Amino-o-cresol, 6-Amino-2-methylphenol
A121702-10G
A121702-1G
A121702-5G
ALA121702-10GEA
229.81
USD
ALA121702-10G
ALA121702-1G
ALA121702-5G
2-Amino-6-methylphenol 92%
2-Amino-6-methylphenol
Formula:
C₇H₉NO MW: 123.1525 g/mol |
MDL Number:
MFCD03788432 CAS Number: 17672-22-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...