4-Amino-3-ethylbenzonitrile ≥96%

Supplier: ALADDIN SCIENTIFIC
Danger

A121620-1G A121620-5G
ALA121620-1GEA 52.16 USD
ALA121620-1G ALA121620-5G
4-Amino-3-ethylbenzonitrile ≥96%
4-Amino-3-ethylbenzonitrile
Formula: C₉H₁₀N₂
Melting Pt: 60-66°C
MDL Number: MFCD00041431
CAS Number: 170230-87-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=682342c52ddc94d55578d276c7fb188cebcb5d3c&vid=682342c52ddc94d55578d276c7fb188cebcb5d3c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR