1-Acetylindoline ≥98%

Supplier: ALADDIN SCIENTIFIC

Synonyms: 1-(Indolin-1-yl)ethanone

A121612-100G A121612-25G A121612-5G
ALA121612-100GEA 497.71 USD
ALA121612-100G ALA121612-25G ALA121612-5G
1-Acetylindoline ≥98%
1-Acetylindoline

It is a tertiary amide having bulky N-groups. Its reduction to the corresponding amine, by reaction with silane in the presence of MoO2Cl2 (catalyst), has been reported.. Reactant for preparation of triazolothiadiazepine dioxide derivatives . Reactant for preparation of halo-substituted aromatic amides . Reactant for preparation of [[(phenyl)piperazinyl]alkyl]indolyl]ethanone and [[[(phenoxyethyl)piperazinyl]alkyl]indolyl]ethanone derivatives as α1-adrenoreceptor antagonists . Reactant for synthesis of naphthalenedione derivatives as antimycobacterial agents . Reactant for regioselective ortho Suzuki-Miyaura coupling reaction It may be used in the synthesis of 5-chloroindole.

Formula: C₁₀H₁₁NO
MW: 161.2
Melting Pt: 102-104°C
MDL Number: MFCD00022908
CAS Number: 16078-30-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6481e0ba46d9962fb23e57efee8674f60faf1ddf&vid=6481e0ba46d9962fb23e57efee8674f60faf1ddf&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR