4-Amino-1-benzylpiperidine ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 4-Amino-N-benzylpiperidine

A121577-100G A121577-5G
ALA121577-100GEA 299.16 USD
ALA121577-100G ALA121577-5G
4-Amino-1-benzylpiperidine ≥98%
4-Amino-1-benzylpiperidine

4-Amino-1-benzylpiperidine was used in the preparation of: butyl 4-amino-1-piperidineacetate, key intermediate required for the synthesis of butyl 4-[(4-amino-5-chloro-2-methoxybenzoyl)amino]-1-piperidineacetate 5-alkylimino-1,2,4-thiadiazolidine-3-ones, such as 4-ethyl-5-[imino-[1-(phenylmethyl)-4-piperidinyl]]-2-methyl-1,2,4-thiadiazolidin-3-one and 4-benzyl-5-[imino-[1-(phenylmethyl)-4-piperidinyl]]-2-isopropyl-1,2,4-thiadiazolidin-3-one glycyrrhetinic acid derivatives

Formula: C₁₂H₁₈N₂
MW: 190.29 g/mol
Density: 0.933 g/cm³
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00006504
CAS Number: 50541-93-0
EINECS: 256-620-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b335573a580cd79f764ee2291c008e01bb611cfb&vid=b335573a580cd79f764ee2291c008e01bb611cfb&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR