4-Amino-1-boc-piperidine ≥97%
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Danger
|
A121575-1G
A121575-25G
A121575-5G
ALA121575-1GEA
39.9
USD
ALA121575-1G
ALA121575-25G
ALA121575-5G
4-Amino-1-boc-piperidine ≥97%
4-Amino-1-Boc-piperidine
Employed in a microwave-assisted solid-phase synthesis of N-substituted piperidines via direct annulation of primary amines with resin-bound dimesylates
Formula:
C₁₀H₂₀N₂O₂ Melting Pt: 45°C Storage Temperature: Ambient |
MDL Number:
MFCD01076201 CAS Number: 87120-72-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...