3-Aminopiperidin-2-one ≥98%
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Danger
|
Synonyms:
3-Amino-2-piperidinone, 3-Amino-2-piperidone, 2-Amino-delta-valerolactam
A121541-1G
A121541-250MG
ALA121541-1GEA
200.36
USD
ALA121541-1G
ALA121541250MG
3-Aminopiperidin-2-one ≥98%
3-Aminopiperidin-2-one
Formula:
C5H10N2O MW: 114.1457 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD06809610 CAS Number: 1892-22-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...