3-Amino-1-Boc-piperidine ≥97%

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 1-BOC-3-Aminopiperidine, (±)-3-Amino-1-boc-piperidine, tert-Butyl 3-aminopiperidine-1-carboxylate

A121536-1G A121536-25G A121536-5G
ALA121536-1GEA 35.06 USD
ALA121536-1G ALA121536-25G ALA121536-5G
3-Amino-1-Boc-piperidine ≥97%
3-Amino-1-Boc-piperidine

Product Application:3-Amino-1-Boc-piperidine is used as an organic chemical synthesis intermediate.

Formula: C₁₀H₂₀N₂O₂
Density: 1.02 g/cm³
Flash Pt: 122 °C
Storage Temperature: Ambient
MDL Number: MFCD01861219
CAS Number: 184637-48-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=58e2933a1a72e62a2bf16008eecb20d2bc715619&vid=58e2933a1a72e62a2bf16008eecb20d2bc715619&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR