(R)-3-Amino-1-benzylpiperidine ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

A121535-1G A121535-5G
ALA121535-1GEA 61.66 USD
ALA121535-1G ALA121535-5G
(R)-3-Amino-1-benzylpiperidine ≥97%
(R)-3-Amino-1-benzylpiperidine

Application:(R)-3-Amino-1-benzylpiperidine is used as an organic chemical synthesis intermediate.

Formula: C₁₂H₁₈N₂
MW: 190.29 g/mol
MDL Number: MFCD03093374
CAS Number: 168466-84-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f595fd933ac4039f703fd8a98efb2dc3ef9a3f91&vid=f595fd933ac4039f703fd8a98efb2dc3ef9a3f91&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR