Piperidin-3-amine dihydrochloride ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: piperidin-3-amine dihydrochloride, 3-Aminopiperidine dihydrochloride, 3-Amino-Piperidine Di-Hydrochloride

A121533-1G A121533-250MG A121533-50MG A121533-5G
ALA121533-1GEA 475.86 USD
ALA121533-1G ALA121533250MG ALA121533-50MG ALA121533-5G
Piperidin-3-amine dihydrochloride ≥97%
Piperidin-3-amine dihydrochloride

Estimation of (S)-piperidin-3-amine in (R)-piperidin-3-amine dihydrochloride (3-aminopiperidine dihydrochloride) has been investigated by chiral high-performance liquid chromatographic (chiral HPLC) methodReactant for N-arylation of heterocyclic diamines Reactant for synthesis of substituted quinolones with reduced phototoxic risk

Formula: C₅H₁₄Cl₂N₂
MW: 173.081 g/mol
Melting Pt: 230°C
Storage Temperature: Ambient
MDL Number: MFCD00012773
CAS Number: 138060-07-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c9d5bafa8a3b38d7e256ace15daacfc2f5588bb&vid=4c9d5bafa8a3b38d7e256ace15daacfc2f5588bb&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR