(S)-(+)-3-Aminopiperidine dihydrochloride ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: (S)-(+)-3-Piperidinamine dihydrochloride, (S)-Piperidin-3-amine dihydrochloride

A121532-250MG A121532-25G A121532-5G A121532-1G
ALA121532250MGEA 23.66 USD
ALA121532250MG ALA121532-25G ALA121532-5G ALA121532-1G
(S)-(+)-3-Aminopiperidine dihydrochloride ≥98%
(S)-(+)-3-Aminopiperidine dihydrochloride
Formula: C₅H₁₄Cl₂N₂
MW: 173.08 g/mol
Melting Pt: 205-209°C
Storage Temperature: Ambient
MDL Number: MFCD03427036
CAS Number: 334618-07-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=46eb06cf7c952cf116fae6536180bf076484a593&vid=46eb06cf7c952cf116fae6536180bf076484a593&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR