2-Amino-9-fluorenone ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A121491-1G A121491-25G A121491-5G
ALA121491-1GEA 36.01 USD
ALA121491-1G ALA121491-25G ALA121491-5G
2-Amino-9-fluorenone ≥98%
2-Amino-9-fluorenone

Application:2-Amino-9-fluorenone is used as an active pharmaceutical intermediate.

Formula: C₁₃H₉NO
Melting Pt: 157-160°C
Storage Temperature: Ambient
MDL Number: MFCD00001157
CAS Number: 3096-57-9
EINECS: 221-445-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd84a1d0f3151c2ce823d61933b072db0c584225&vid=dd84a1d0f3151c2ce823d61933b072db0c584225&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR