1-Aminoanthracene ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 1-Anthramine, Anthracen-1-amine

A121482-1G A121482-250MG
ALA121482-1GEA 112.01 USD
ALA121482-1G ALA121482250MG
1-Aminoanthracene ≥98%
1-Aminoanthracene
Formula: C₁₄H₁₁N
MW: 193.25 g/mol
Melting Pt: 119.0 - 123.0 °C
MDL Number: MFCD00003579
CAS Number: 610-49-1
EINECS: 210-225-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c34eb834504439bb0efc3e28eeb548a728bbbe4e&vid=c34eb834504439bb0efc3e28eeb548a728bbbe4e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR