4-Amino-p-terphenyl ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A121452-1G A121452-5G
ALA121452-1GEA 73.06 USD
ALA121452-1G ALA121452-5G
4-Amino-p-terphenyl ≥97%
4-Amino-p-terphenyl
Formula: C₁₈H₁₅N
Melting Pt: 201 °C
Storage Temperature: Ambient
MDL Number: MFCD00191302
CAS Number: 7293-45-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=41cd6fad0fe3d50e7d1f46c512ee7c485a80291f&vid=41cd6fad0fe3d50e7d1f46c512ee7c485a80291f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR