L-Alanyl-L-tyrosine ≥98% (by HPLC)

Supplier: ALADDIN SCIENTIFIC

Synonyms: H-Ala-Tyr-OH, (S)-2-((S)-2-Aminopropanamido)-3-(4-hydroxyphenyl)propanoic acid, L-Alanyl-L-tyrosine

A121379-1G A121379-25G A121379-5G
ALA121379-1GEA 162.36 USD
ALA121379-1G ALA121379-25G ALA121379-5G
L-Alanyl-L-tyrosine ≥98% (by HPLC)
L-Alanyl-L-tyrosine
Formula: C₁₂H₁₆N₂O₄
MW: 252.27 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00038164
CAS Number: 3061-88-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fbfe89e20c6885a3691c82ebcfd12d20f8957ecd&vid=fbfe89e20c6885a3691c82ebcfd12d20f8957ecd&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR