4-Amino-3-chlorophenol hydrochloride ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 4-Amino-3-chlorophenol Hydrochloride, 2-Chloro-4-hydroxyaniline hydrochloride

A121105-100G A121105-1G A121105-25G A121105-5G
ALA121105-100GEA 349.51 USD
ALA121105-100G ALA121105-1G ALA121105-25G ALA121105-5G
4-Amino-3-chlorophenol hydrochloride ≥98%
4-Amino-3-chlorophenol hydrochloride
Formula: C₆H₇Cl₂NO
Melting Pt: 254-256°C
MDL Number: MFCD00143110
CAS Number: 52671-64-4
EINECS: 610-882-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee88bf152d2b77279bf31868c0406ca82eb301e1&vid=ee88bf152d2b77279bf31868c0406ca82eb301e1&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR