Amylose (from potato) amylase substrate
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
A120987-1G
A120987-250MG
ALA120987-1GEA
113.91
USD
ALA120987-1G
ALA120987250MG
Amylose (from potato) amylase substrate
Amylose
Amylose is a polyglucose polysaccharide which together with amylopectin makes starch(s). The glucosidic bonds of amylopectin are α-1,4 linkages. Amylose is used to identify, differentiate and characterize amylase(s). Amylose is used as a thickening agent, a gelling agent and emulsion stabilizer. Amylose is used in the preparation of films and membranes with potential use in drug A substrate for serum amylase.Application:
Amylose in potato has been used as a standard to determine the apparent amylose content (AAC) of rice starch by iodine-binding colorimetry.
Formula:
(C₆H₁₀O₅)n |
MDL Number:
MFCD00134652 CAS Number: 9005-82-7 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...