4-Amino-3-fluorobenzoic acid ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 4-Amino-3-fluorobenzoic Acid

A120972-100G A120972-1G A120972-250MG
ALA120972-100GEA 381.81 USD
ALA120972-100G ALA120972-1G ALA120972250MG
4-Amino-3-fluorobenzoic acid ≥98%
4-Amino-3-fluorobenzoic acid
Formula: C₇H₆FNO₂
MW: 155.13 g/mol
Melting Pt: 214-216°C
Storage Temperature: Ambient
MDL Number: MFCD01660374
CAS Number: 455-87-8
EINECS: 207-250-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8f61e8065abe9eaa2134cff492165b734b9a9a3a&vid=8f61e8065abe9eaa2134cff492165b734b9a9a3a&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR